missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bavituximab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Bavituximab Chimeric
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284254
GREENER_CHOICE
View Documents
Invitrogen™
MA559747
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283935
View Documents
Invitrogen™
MA559748
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Bavituximab is a chimeric Anti-PS monoclonal antibody analog which is used to potentially treat cancers and viral infections. It binds to phosphatidylserine and other exposed host cell lipids when induced by cellular stress. Additional analogs in the class include 3G4, 2aG4, 9d2 and Hu3g4. Bavituximab Anti-Cancer is a monoclonal antibody that binds to a basic component of the cell structure called a phospholipid that is exposed only on the surface of tumor blood vessel cells or on cells infected with certain viruses. Bavituximab binding to the tumor blood vessel cells alerts the body's immune system to attack the tumor and its blood supply. This has been shown to inhibit tumor growth and development. Because in healthy cells the phospholipids are concealed inside the cell, the bavituximab does not bind to them. This targets the bavituximab to the malignant cells and potentially minimizes unwanted side effects.
Specifications

Specifications

Bavituximab Chimeric
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Tarvacin ch3G4
Antibody
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.