missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bavituximab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559748
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Bavituximab is a chimeric Anti-PS monoclonal antibody analog which is used to potentially treat cancers and viral infections. It binds to phosphatidylserine and other exposed host cell lipids when induced by cellular stress. Additional analogs in the class include 3G4, 2aG4, 9d2 and Hu3g4. Bavituximab Anti-Cancer is a monoclonal antibody that binds to a basic component of the cell structure called a phospholipid that is exposed only on the surface of tumor blood vessel cells or on cells infected with certain viruses. Bavituximab binding to the tumor blood vessel cells alerts the body's immune system to attack the tumor and its blood supply. This has been shown to inhibit tumor growth and development. Because in healthy cells the phospholipids are concealed inside the cell, the bavituximab does not bind to them. This targets the bavituximab to the malignant cells and potentially minimizes unwanted side effects.
Specifications
| Bavituximab Chimeric | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Tarvacin ch3G4 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction