missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Batoclimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559106
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Batoclimab (HBM9161) is a fully human antibody targeting the neonatal Fc receptor (FcRn). It targets patients with neuromyelitis optica spectrum diseases and other autoimmune diseases, and can accelerate the degradation of autoantibodies in their bodies. Batoclimab (HBM9161) is an antibody that targets the neonatal Fc receptor (FcRn). FcRn expression is up-regulated by pro-inflammatory cytokines (such as TNF-alpha) and prolongs the half-life of IgG and serum albumin by reducing lysosomal degradation in endothelial cells and bone marrow-derived cells. Blocking the FcRn-IgG interaction can accelerate the degradation of autoantibodies and reduce the onset of various pathogenic IgG-mediated autoimmune diseases, including myasthenia gravis, thyroid ophthalmopathy, neuromyelitis optica spectrum disease and immunity Thrombocytopenia.
Specifications
| Batoclimab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HL161 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 λ2 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction