missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAT5 Antibody (S01-4D1), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | BAT5 |
|---|---|
| Clone | S01-4D1 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643854
|
Novus Biologicals
NBP3-19580-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663835
|
Novus Biologicals
NBP3-19580-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAT5 Monoclonal antibody specifically detects BAT5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| BAT5 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 7920 | |
| IgG | |
| Affinity purified |
| S01-4D1 | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| abhydrolase domain containing 16A, BAT5G5, D6S82EEC 3.-, HLA-B associated transcript 5, HLA-B-associated transcript 5, NG26protein BAT5, PP199, Protein G5 | |
| A synthetic peptide of human BAT5 (Uniprot # O95870) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title