missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAT5 Antibody (S01-4D1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19580-50ul
This item is not returnable.
View return policy
Description
BAT5 Monoclonal antibody specifically detects BAT5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| BAT5 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7920 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| S01-4D1 | |
| Unconjugated | |
| abhydrolase domain containing 16A, BAT5G5, D6S82EEC 3.-, HLA-B associated transcript 5, HLA-B-associated transcript 5, NG26protein BAT5, PP199, Protein G5 | |
| A synthetic peptide of human BAT5 (Uniprot # O95870) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction