missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Bastotest™ (CE/IVD)
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
- This reagent kit allows the quantitative determination of basophil degranulation in heparinized human whole blood.
- Contains the chemotactic peptide N-formyl-Met-Leu-Phe (fMLP) as positive control, important allergens, a two-color antibody reagent for assessing the activation of human basophilic granulocytes and necessary reagents.
- Determines the percentage of basophilic granulocytes that have degranulated after incubation with allergen or fMLP.
Specifications
Specifications
| Content And Storage | Store the kit in the dark at 2 - 8°C (in refrigerator). The fMLP and allergen working solutions have to be dicarded after use. Reagent B (stimulation buffer) has to be stored in aliquots at <−15°C after reconstitution. The reagents are supplied sterile with a preservative that does not influence activation of basophilic granulocytes. The reagents are stable for the period shown on the packaging label, when stored as described |
| Detection Method | Flow Cytometry, Fluorochrome |
| Includes | Instamed Salts, Stimulation Buffer, Mite Mix Allergenic Extract, 7 Grass Mix Allergenic Extract, fMLP Chemotactic Peptide, anti-IgE-PE, anti-CD63-FITC, Lysing Solution |
| Quantity | 100 Tests |
| Sample Type | Heparinized Human Whole Blood |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction