missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bassoon Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Bassoon |
|---|---|
| Concentration | 550 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18088105
|
Proteintech
30392-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18087115
|
Proteintech
30392-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Bassoon antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Bassoon. This antibody recognizes human, mouse antigen. The Bassoon antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Bassoon | |
| Immunohistochemistry, ELISA | |
| Rabbit | |
| 8927 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 550 μg/mL | |
| Liquid | |
| Human, Mouse | |
| BSN, KIAA0434, Protein bassoon, Zinc finger protein 231, ZNF231 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title