Learn More
Bassoon Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30392-1-AP-150UL
Description
The Bassoon antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Bassoon. This antibody recognizes human, mouse antigen. The Bassoon antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Bassoon | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| BSN, KIAA0434, Protein bassoon, Zinc finger protein 231, ZNF231 | |
| 150 μL | |
| 8927 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |