missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BASP1 Monoclonal Antibody (K9N006_12F10), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | BASP1 |
|---|---|
| Clone | K9N006_12F10 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30210722
|
Invitrogen™
MA555861 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30212559
|
Invitrogen™
MA556241 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GYNVNDEKAK EKDKKAEGAA TEEEGTPKES EPQAAAEPAE AKEGKEKPDQ DAEGKAEEKE GEKDAAAAKE EAPKAEPEKT EGAAEAKAEP PKAPEQEQAA PGPAAGGEAP KAAEAAAAPA ESAAPAAGEE PSKEEGEPKK
BASP1 Monoclonal antibody specifically detects BASP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| BASP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 22 kDa neuronal tissue-enriched acidic protein; 2610024P12Rik; BASP1; brain abundant membrane attached signal protein 1; brain abundant, membrane attached signal protein 1; brain acid soluble protein 1; brain acidic membrane protein; brain acid-soluble protein 1; CAP23; CAP-23; Ckap3; cytoskeleton associated protein 3; MGC8555; NAP22; NAP-22; neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein | |
| Recombinant human BASP1 (immunogen range is 11-150). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| BASP1 |
| K9N006_12F10 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P80723 | |
| 10409 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title