missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BASP1 Monoclonal Antibody (K9N006_12F10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556241
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: GYNVNDEKAK EKDKKAEGAA TEEEGTPKES EPQAAAEPAE AKEGKEKPDQ DAEGKAEEKE GEKDAAAAKE EAPKAEPEKT EGAAEAKAEP PKAPEQEQAA PGPAAGGEAP KAAEAAAAPA ESAAPAAGEE PSKEEGEPKK
BASP1 (brain acid soluble protein 1) is a membrane bound protein with several transient phosphorylation sites and PEST motifs. Conservation of proteins with PEST sequences among different species supports their functional significance. PEST sequences typically occur in proteins with high turnover rates. Immunological characteristics of this protein are species specific. This protein also undergoes N-terminal myristoylation.
Specifications
| BASP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P80723 | |
| BASP1 | |
| Recombinant human BASP1 (immunogen range is 11-150). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| K9N006_12F10 | |
| Unconjugated | |
| BASP1 | |
| 22 kDa neuronal tissue-enriched acidic protein; 2610024P12Rik; BASP1; brain abundant membrane attached signal protein 1; brain abundant, membrane attached signal protein 1; brain acid soluble protein 1; brain acidic membrane protein; brain acid-soluble protein 1; CAP23; CAP-23; Ckap3; cytoskeleton associated protein 3; MGC8555; NAP22; NAP-22; neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 10409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction