missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barnstead™ Classic Stills Storage Tanks

Ensure the purity of distilled water with the copper and tin-crafted reservoirs of Thermo Scientific™ Barnstead™ Classic Stills Storage Tanks.

£3764.06 - £5137.11

Specifications

For Use With (Equipment) Classic Still
Type Storage Tank
View More Specs

Products 2
Product Code Brand Capacity (English) Price Quantity & Availability  
Product Code Brand Capacity (English) Price Quantity & Availability  
13478239
View Documents Promotion Details
Thermo Scientific™
B3043
10 gal. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
13438249
View Documents Promotion Details
Thermo Scientific™
B3045
25 gal.
£5137.11
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
 
Description

Description

Ensure the purity of distilled water with the copper and tin-crafted reservoirs of Thermo Scientific™ Barnstead™ Classic Stills Storage Tanks. Select from 10 to 200 gallon reservoir options. All reservoirs include a removable cover, water level sight glass and a tin-coated draw-off faucet with serrated hose nipple. Connections allow for optional accessories. Tanks require a floor stand or wall bracket (10 and 25 gallon reservoirs only) for proper installation.

Ordering Information:

Replacement consumables include UV Lamp (04141), Ventgard Air Filter (H3120), replacement Water Seal (H3130) and Ventgard Filter Element (25001); all sold separately.

Requires:

Floor stand (all sizes) or wall bracket (10- and 25 gal. reservoirs only) are required for secure installation. Ventgard Air Filter/Water Seal (H3111), and UV Light Assembly (H4005) are also required to protect pure water during storage.

Specifications

Specifications

Classic Still
Storage Tank
Videos
SDS
Documents

Documents

Product Certifications