missing translation for 'onlineSavingsMsg'
Learn More

Barnidipine Hydrochloride, TRC

Product Code. 30137400
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137400 100 mg 100mg
30134263 25 mg 25mg
30012849 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137400 Supplier Toronto Research Chemicals Supplier No. TRCB118700100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104757-53-1
Molecular Formula C27 H29 N3 O6 . Cl H
Molecular Weight (g/mol) 528
Synonym 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 3-methyl 5-[(3S)-1-(phenylmethyl)-3-pyrrolidinyl] ester, hydrochloride (1:1), (4S)-, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, methyl (3S)-1-(phenylmethyl)-3-pyrrolidinyl ester, monohydrochloride, (4S)- (9CI), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, methyl 1-(phenylmethyl)-3-pyrrolidinyl ester, monohydrochloride, [S-(R*,R*)]-, Barnidipine hydrochloride, YM 09730-5
IUPAC Name 5-O-[(3S)-1-benzylpyrrolidin-3-yl] 3-O-methyl (4S)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
SMILES Cl.COC(=O)C1=C(C)NC(=C([C@H]1c2cccc(c2)[N+](=O)[O-])C(=O)O[C@H]3CCN(Cc4ccccc4)C3)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C27H29N3O6.ClH/c1-17-23(26(31)35-3)25(20-10-7-11-21(14-20)30(33)34)24(18(2)28-17)27(32)36-22-12-13-29(16-22)15-19-8-5-4-6-9-19;/h4-11,14,22,25,28H,12-13,15-16H2,1-3H3;1H/t22-,25-;/m0./s1
Formula Weight 527.1823
Chemical Name or Material Barnidipine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.