missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BARHL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £368.00
Specifications
| Antigen | BARHL2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18394475
|
Novus Biologicals
NBP3-10922-25UL |
25 μg |
£161.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18306665
|
Novus Biologicals
NBP3-10922-100UL |
100 μg |
£368.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BARHL2 Polyclonal specifically detects BARHL2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| BARHL2 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BarH (Drosophila)-like 2, barH-like 2 homeobox protein, BarH-like homeobox 2 | |
| The immunogen is a synthetic peptide directed towards the middle region of human BARHL2 (NP_064447). Peptide sequence SSPHHTPKQESNAVHESFRPKLEQEDSKTKLDKREDSQSDIKCHGTKEEG | |
| Affinity purified |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 343472 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title