missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BARHL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-10922-100UL
This item is not returnable.
View return policy
Description
BARHL2 Polyclonal specifically detects BARHL2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BARHL2 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| BarH (Drosophila)-like 2, barH-like 2 homeobox protein, BarH-like homeobox 2 | |
| The immunogen is a synthetic peptide directed towards the middle region of human BARHL2 (NP_064447). Peptide sequence SSPHHTPKQESNAVHESFRPKLEQEDSKTKLDKREDSQSDIKCHGTKEEG | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 343472 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction