missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BARD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | BARD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
BARD1 Polyclonal antibody specifically detects BARD1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| BARD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer | |
| PBS with 50% glycerol, pH7.3. | |
| 580 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BARD-1, BRCA1 associated RING domain 1, BRCA1-associated RING domain gene 1, BRCA1-associated RING domain protein 1, EC 6.3.2.- | |
| Recombinant protein of mouse BARD1 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title