missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BARD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92343-0.02ml
This item is not returnable.
View return policy
Description
BARD1 Polyclonal antibody specifically detects BARD1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BARD1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BARD-1, BRCA1 associated RING domain 1, BRCA1-associated RING domain gene 1, BRCA1-associated RING domain protein 1, EC 6.3.2.- | |
| Recombinant protein of mouse BARD1 | |
| 0.02 mL | |
| Breast Cancer, Cancer | |
| 580 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction