Learn More
Agilent BioTek Barcode Scanner Kit
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Barcode Labeled Plates; Biostack; LHC Secure software for 405TS/LS, EL406 and MultiFlo FX; Gen5 Software for other Agilent BioTek Microplate Readers, Washers and Dispensers
Unit Size:
Each
Description
- 405TS/LS , EL406 and MultiFlo FX must be under computer control via LHC Secure software (sold separately) in order to work with barcode labeled plates and BioStack barcode scanner
Specifications
Specifications
| Product Type | Barcode Scanner Kit |
| For Use With (Equipment) | Barcode Labeled Plates; Biostack; LHC Secure software for 405TS/LS, EL406 and MultiFlo FX; Gen5 Software for other Agilent BioTek Microplate Readers, Washers and Dispensers |