missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP29 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76421-0.025mg
This item is not returnable.
View return policy
Description
BAP29 Polyclonal specifically detects BAP29 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| BAP29 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Bap29, BAP29DKFZp686M2086, B-cell receptor-associated protein 29, BCR-associated protein 29, FLJ53907 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| NP_001008405 | |
| BCAP29 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the center of human Bap29. Bap29 antibody is specific for isoform a of Bap29. The immunogen is located within amino acids 230 - 280 of BAP29. Amino Acid Squence: KRSHKNGSIGKQTGSRK | |
| 0.025 mg | |
| Apoptosis | |
| 55973 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction