missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP29 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | BAP29 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
BAP29 Polyclonal specifically detects BAP29 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| BAP29 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS with 0.02% Sodium Azide | |
| Bap29, BAP29DKFZp686M2086, B-cell receptor-associated protein 29, BCR-associated protein 29, FLJ53907 | |
| BCAP29 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_001008405 | |
| 55973 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the center of human Bap29. Bap29 antibody is specific for isoform a of Bap29. The immunogen is located within amino acids 230 - 280 of BAP29. Amino Acid Squence: KRSHKNGSIGKQTGSRK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title