missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BANK1 Monoclonal Antibody (K92023_11B1), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | BANK1 |
|---|---|
| Clone | K92023_11B1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30206167
|
Invitrogen™
MA555882 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30204165
|
Invitrogen™
MA556262 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: PGTMVEGQME RSQNWGHPGV RQETGDEPKG EKEKKEEEKE QEEEEDPYTF AEIDDSEYDM ILANLSIKKK TGSRSFIINR PPAPTPRPTS IPPKEETTPY IAQVFQQKTA RRQSDDDKFC GLPKKQDRAR IESPAFSTLR GCLTDGQEEL ILLQEKVKNG
BANK1 Monoclonal antibody specifically detects BANK1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| BANK1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| A530094C12Rik; AI451642; AVIEF; B cell scaffold protein with ankyrin repeats 1; BANK; Bank1; B-cell scaffold protein with ankyrin repeats; B-cell scaffold protein with ankyrin repeats 1; B-cell scaffold protein with ankyrin repeats 1 delta 2 transcript; LRRGT00104; Protein AVIEF | |
| Recombinant human BANK1 (immunogen range is 531-690). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| BANK1 |
| K92023_11B1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8NDB2 | |
| 55024 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title