missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BANK1 Monoclonal Antibody (K92023_11B1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556262
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: PGTMVEGQME RSQNWGHPGV RQETGDEPKG EKEKKEEEKE QEEEEDPYTF AEIDDSEYDM ILANLSIKKK TGSRSFIINR PPAPTPRPTS IPPKEETTPY IAQVFQQKTA RRQSDDDKFC GLPKKQDRAR IESPAFSTLR GCLTDGQEEL ILLQEKVKNG
The protein encoded by this gene is a B-cell-specific scaffold protein that functions in B-cell receptor-induced calcium mobilization from intracellular stores. This protein can also promote Lyn-mediated tyrosine phosphorylation of inositol 1,4,5-trisphosphate receptors. Polymorphisms in this gene are associated with susceptibility to systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
Specifications
| BANK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8NDB2 | |
| BANK1 | |
| Recombinant human BANK1 (immunogen range is 531-690). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K92023_11B1 | |
| Unconjugated | |
| BANK1 | |
| A530094C12Rik; AI451642; AVIEF; B cell scaffold protein with ankyrin repeats 1; BANK; Bank1; B-cell scaffold protein with ankyrin repeats; B-cell scaffold protein with ankyrin repeats 1; B-cell scaffold protein with ankyrin repeats 1 delta 2 transcript; LRRGT00104; Protein AVIEF | |
| Mouse | |
| Protein A | |
| RUO | |
| 55024 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction