missing translation for 'onlineSavingsMsg'
Learn More

Baicalin, MedChemExpress

Product Code. 16499026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499026 100 mg 100mg
16492509 1 g 1g
16481328 5 g 5g
16452366 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499026 Supplier Medchem Express Supplier No. HYN0197
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Baicalin, as a flavonoid glycoside, is an allosteric carnitine palmityl transferase 1 (CPT1) activator. Baicalin reduces the expression of NF-κB.

Baicalin, as a flavonoid glycoside, is an allosteric carnitine palmityl transferase 1 (CPT1) activator. Baicalin reduces the expression of NF-κB.

Specifications

Chemical Name or Material Baicalin
CAS 21967-41-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H18O11
Quantity 100 mg
Synonym Baicalein 7-O-β-D-glucuronide
Solubility Information DMSO : ≥ 100 mg/mL (224.03 mM) ∣H2O : 0.1 mg/mL (0.22 mM; Need ultrasonic)
SMILES O[C@H]([C@H]([C@@H]([C@@H](C(O)=O)O1)O)O)[C@@H]1OC2=C(O)C(O)=C3C(C=C(C4=CC=CC=C4)OC3=C2)=O
Molecular Weight (g/mol) 446.36
Formula Weight 446.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.