missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
£183.00 - £423.00
Specifications
| Antigen | BAI1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot reported in scientific literature, Immunohistochemistry 1:1000-1:3000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000-1:3000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18229880
|
Novus Biologicals
NB110-81586SS |
0.025 mL |
£183.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18720194
|
Novus Biologicals
NB110-81586 |
0.1 mL |
£423.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAI1 Polyclonal specifically detects BAI1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| BAI1 | |
| Western Blot reported in scientific literature, Immunohistochemistry 1:1000-1:3000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000-1:3000 | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Angiogenesis, Apoptosis, Cancer, GPCR, Neurodegeneration, Neuroscience | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
| BAI1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| O14514 | |
| 575 | |
| Synthetic peptide made to a C-terminal portion of the human BAI1 protein (within residues 1500-1584). [Swiss-Prot# O14514] | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title