missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB110-81586
This item is not returnable.
View return policy
Description
BAI1 Polyclonal specifically detects BAI1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| BAI1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot reported in scientific literature, Immunohistochemistry 1:1000-1:3000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000-1:3000 | |
| O14514 | |
| BAI1 | |
| Synthetic peptide made to a C-terminal portion of the human BAI1 protein (within residues 1500-1584). [Swiss-Prot# O14514] | |
| 0.1 mL | |
| Alzheimers Research, Angiogenesis, Apoptosis, Cancer, GPCR, Neurodegeneration, Neuroscience | |
| 575 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction