missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ BagFilter™ 3500P Lateral Pipette Filter Bag

Product Code. 10736941
Change view
Click to view available options
Capacity (Metric):
3500 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10736941 3500 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10736941 Supplier Fisherbrand™ Supplier No. 113510
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A sterile bag with a side non-woven filter, ideal for pipetting applications in the laboratory.

  • Lateral non-woven filter for all microbiological analyses
  • Flora is extracted from the sample and goes through the filter during blending
  • Ideal for homogenizing fibrous samples
  • Multilayer, reinforced multicoated complex
  • Porosity of the filter is less than 250 microns
  • Rigid and transparent
  • No need to wait for debris sedimentation
  • Particle-free solution with precise results and excellent reproducibility
  • Gamma ray treated
  • Same bag is used to homogenize, filter and pipet
  • Can be closed with a clip or heat-sealed
  • Compatible with any lab blender
  • Approved for food contact

Specifications

Closure Type Heat Seal or Clip
Color Transparent
For Use With (Application) Microbiological analyses
Sterility Sterile
No. per Pack 10
Permeability Less than 250 microns
Quantity Pack of 10; Case of 100
Sterilization Method Gamma Irradiated
Dimensions (L x W) 380 x 600mm
Product Type Lateral Filter Bag
Capacity (Metric) 3500 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.