missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£180.00 - £291.00
Specifications
| Antigen | BAG5 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18077565
|
Novus Biologicals
NB100-56091SS |
0.025 mL |
£180.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18779223
|
Novus Biologicals
NB100-56091 |
0.05 mL |
£291.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAG5 Polyclonal specifically detects BAG5 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| BAG5 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Bovine, Canine | |
| BAG family molecular chaperone regulator 5, BAG-5, BAG-family molecular chaperone regulator-5, Bcl-2-associated athanogene 5, BCL2-associated athanogene 5, KIAA0873 | |
| BAG5 | |
| IgG | |
| Unpurified | |
| BAG5 |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antisera with 0.05% Sodium Azide | |
| 9529 | |
| A synthetic peptide corresponding to amino acids 168-187 (KKQPSLPLSEDAHPSVAKIN) of human BAG-5 was used as immunogen, GenBank no. gi 6631077 ref NP_004864.1 . The amino acid sequence used for immunogen is 100% consevered with chicken and dog BAG-5, 95% conserved with mouse BAG-5, and 94% conserved with rat BAG-5. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title