missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-56091SS
This item is not returnable.
View return policy
Description
BAG5 Polyclonal specifically detects BAG5 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| BAG5 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| BAG family molecular chaperone regulator 5, BAG-5, BAG-family molecular chaperone regulator-5, Bcl-2-associated athanogene 5, BCL2-associated athanogene 5, KIAA0873 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 9529 | |
| Human, Mouse, Rat, Bovine, Canine | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| BAG5 | |
| A synthetic peptide corresponding to amino acids 168-187 (KKQPSLPLSEDAHPSVAKIN) of human BAG-5 was used as immunogen, GenBank no. gi 6631077 ref NP_004864.1 . The amino acid sequence used for immunogen is 100% consevered with chicken and dog BAG-5, 95% conserved with mouse BAG-5, and 94% conserved with rat BAG-5. | |
| 0.025 mL | |
| Primary | |
| BAG5 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction