missing translation for 'onlineSavingsMsg'
Learn More

BAG3 Rabbit anti-Human, Mouse, Clone: 8P3W7, Novus Biologicals™

Rabbit Monoclonal Antibody

£150.00 - £386.00

Specifications

Antigen BAG3
Clone 8P3W7
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18366824
View Documents
Novus Biologicals
NBP3-16469-20UL
20 μg
£150.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18378855
View Documents
Novus Biologicals
NBP3-16469-100UL
100 μg
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BAG3 Monoclonal antibody specifically detects BAG3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

BAG3
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse
BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human BAG3 (O95817).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
8P3W7
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Cancer, Cell Biology, Endocrinology
PBS, 0.05% BSA, 50% glycerol, pH7.3
9531
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.