missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bag-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£168.00 - £242.00
Specifications
| Antigen | Bag-1 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Simple Western 1:2500, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18024185
|
Novus Biologicals
NB100-56086SS |
0.025 mL |
£168.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18797603
|
Novus Biologicals
NB100-56086 |
0.05 mL |
£242.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bag-1 Polyclonal specifically detects Bag-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.Specifications
| Bag-1 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| Whole antisera with 0.02% Sodium Azide | |
| 573 | |
| Full-length recombinant human BAG-1L protein was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Simple Western 1:2500, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| BAG family molecular chaperone regulator 1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 | |
| BAG1 | |
| IgG | |
| Unpurified | |
| BAG1 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title