missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bag-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56086
This item is not returnable.
View return policy
Description
Bag-1 Polyclonal specifically detects Bag-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Specifications
| Bag-1 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Simple Western 1:2500, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| BAG family molecular chaperone regulator 1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| BAG1 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| BAG1 | |
| Full-length recombinant human BAG-1L protein was used as immunogen. | |
| 0.05 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 573 | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction