missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAF57 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£177.00 - £367.00
Specifications
| Antigen | BAF57 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:20-1:50, Knockout Validated |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation, Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647261
|
Novus Biologicals
NBP2-92966-0.02ml |
0.02 mL |
£177.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637280
|
Novus Biologicals
NBP2-92966-0.1ml |
0.1 mL |
£367.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAF57 Polyclonal antibody specifically detects BAF57 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Gene Knock-OutSpecifications
| BAF57 | |
| Western Blot, Immunofluorescence, Immunoprecipitation, Gene Knock-Out | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS with 50% glycerol, pH7.3. | |
| 6605 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:20-1:50, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BAF57BRG1-associated factor 57, chromatin remodeling complex BRG1-associated factor 57, subfamily e, member 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin e1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily E member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-411 of human BAF57 (NP_003070.3). MSKRPSYAPPPTPAPATQMPSTPGFVGYNPYSHLAYNNYRLGGNPGTNSRVTASSGITIPKPPKPPDKPLMPYMRYSRKVWDQVKASNPDLKLWEIGKIIGGMWRDLTDEEKQEYLNEYEAEKIEYNESMKAYHNSPAYLAYINAKSRAEAALEEESRQRQSRMEKGEPYMSIQPAEDPDDYDDGFSMKHTATARFQRNHRLISEILSESVVPDVRSVVTTARMQVLKRQVQSLMVHQRKLEAELLQIEERHQEKKRKFLESTDSFNNELKRLCGLKVEVDMEKIAAEIAQAEEQARKRQEEREKEAAEQAERSQSSIVPEEEQAANKGEEKKDDENIPMETEETHLEETTESQQNGEEGTSTPEDKESGQEGVDSMAEEGTSDSNTGSESNSATVEEPPTDPIPEDEKKE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title