missing translation for 'onlineSavingsMsg'
Learn More

BAF57 Antibody - BSA Free, Novus Biologicals™

Product Code. 18647261 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18647261 0.02 mL 0.02mL
18637280 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18647261 Supplier Novus Biologicals Supplier No. NBP2929660.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BAF57 Polyclonal antibody specifically detects BAF57 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAF57
Applications Western Blot, Immunofluorescence, Immunoprecipitation, Gene Knock-Out
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:20-1:50, Knockout Validated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BAF57BRG1-associated factor 57, chromatin remodeling complex BRG1-associated factor 57, subfamily e, member 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin e1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily E member 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-411 of human BAF57 (NP_003070.3). MSKRPSYAPPPTPAPATQMPSTPGFVGYNPYSHLAYNNYRLGGNPGTNSRVTASSGITIPKPPKPPDKPLMPYMRYSRKVWDQVKASNPDLKLWEIGKIIGGMWRDLTDEEKQEYLNEYEAEKIEYNESMKAYHNSPAYLAYINAKSRAEAALEEESRQRQSRMEKGEPYMSIQPAEDPDDYDDGFSMKHTATARFQRNHRLISEILSESVVPDVRSVVTTARMQVLKRQVQSLMVHQRKLEAELLQIEERHQEKKRKFLESTDSFNNELKRLCGLKVEVDMEKIAAEIAQAEEQARKRQEEREKEAAEQAERSQSSIVPEEEQAANKGEEKKDDENIPMETEETHLEETTESQQNGEEGTSTPEDKESGQEGVDSMAEEGTSDSNTGSESNSATVEEPPTDPIPEDEKKE
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6605
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.