missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baculovirus Envelope gp64 protein Antibody (M001), Novus Biologicals™
Mouse Monoclonal Antibody
£308.00 - £392.00
Specifications
| Antigen | Baculovirus Envelope gp64 protein |
|---|---|
| Clone | M001 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626956
|
Novus Biologicals
NBP3-05840-200ug |
200 μg |
£308.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630977
|
Novus Biologicals
NBP3-05840-500ug |
500 μg |
£392.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Baculovirus Envelope gp64 protein Monoclonal antibody specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for NeutralizationSpecifications
| Baculovirus Envelope gp64 protein | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Virus | |
| ACNVgp129, AcOrf-128 | |
| IgG1 | |
| Protein A purified |
| M001 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| Produced in mouse immunized with recombinant baculovirus (Autographa californica multicapsid nucleopolyhedrovirus - AcmNPV) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title