missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baculovirus Envelope gp64 protein Antibody (M001), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05840-200ug
This item is not returnable.
View return policy
Description
Baculovirus Envelope gp64 protein Monoclonal antibody specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Neutralization
Specifications
| Baculovirus Envelope gp64 protein | |
| Monoclonal | |
| Unconjugated | |
| ACNVgp129, AcOrf-128 | |
| Produced in mouse immunized with recombinant baculovirus (Autographa californica multicapsid nucleopolyhedrovirus - AcmNPV) | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Neutralization | |
| M001 | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction