Learn More
BACH1 Rabbit, anti-Human, Clone: 4K14, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82842-7-RR-20UL
Description
The BACH1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human BACH1. This antibody recognizes human antigen. The BACH1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BACH1 | |
| 4K14 | |
| BACH 1, BACH1, BTB and CNC homolog 1, HA2303 | |
| 20 μL | |
| 571 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |