missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACH1 Rabbit, anti-Human, Clone: 4K14, Proteintech
Rabbit Recombinant Monoclonal Antibody
£135.00 - £360.00
Specifications
| Antigen | BACH1 |
|---|---|
| Clone | 4K14 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18004725
|
Proteintech
82842-7-RR-100UL |
100 μL |
£360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020746
|
Proteintech
82842-7-RR-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BACH1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human BACH1. This antibody recognizes human antigen. The BACH1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BACH1 | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| BACH 1, BACH1, BTB and CNC homolog 1, HA2303 | |
| Primary |
| 4K14 | |
| Western Blot, ELISA | |
| Rabbit | |
| 571 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title