missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAAT1 Recombinant Rabbit Monoclonal Antibody (23GB4355), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | BAAT1 |
|---|---|
| Clone | 23GB4355 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088518
![]() |
Invitrogen™
MA552696 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087832
|
Invitrogen™
MA552697 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAAT1 Recombinant Monoclonal antibody specifically detects BAAT1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| BAAT1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q6PJG6 | |
| 221927 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4355 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA881470; BAAT1; Brat1; Brat1-ps1; BRCA1 associated ATM activator 1; BRCA1-associated ATM activator 1; BRCA1-associated ATM activator 1, pseudogene 1; BRCA1-associated protein required for ATM activation protein 1; C7orf27; HEAT repeat-containing protein C7orf27; MGC22916; RGD1565146; RMFSL | |
| A synthesized peptide derived from human BRAT1 (701-821AA). | |
| Primary | |
| -20°C | |
| BRAT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title