missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAAT1 Recombinant Rabbit Monoclonal Antibody (23GB4355), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552696
This item is not returnable.
View return policy
Description
Required for activation of ATM following ionizing radiation. May act by regulating dephosphorylation of ATM.
Specifications
| BAAT1 | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6PJG6 | |
| BRAT1 | |
| A synthesized peptide derived from human BRAT1 (701-821AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4355 | |
| Unconjugated | |
| BRAT1 | |
| AA881470; BAAT1; Brat1; Brat1-ps1; BRCA1 associated ATM activator 1; BRCA1-associated ATM activator 1; BRCA1-associated ATM activator 1, pseudogene 1; BRCA1-associated protein required for ATM activation protein 1; C7orf27; HEAT repeat-containing protein C7orf27; MGC22916; RGD1565146; RMFSL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 221927 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction