Learn More
Invitrogen™ B7-H7 Monoclonal Antibody (MA57YW), PE-Cyanine7, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£169.00 - £383.00
Specifications
| Antigen | B7-H7 |
|---|---|
| Clone | MA57YW |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16380784
|
Invitrogen™
25-6537-41 |
25 Tests |
£169.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16390784
![]() |
Invitrogen™
25-6537-42 |
100 Tests |
£383.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This MA57YW monoclonal antibody recognizes human B7-H7 on live and paraformaldehyde-fixed cells. Applications Reported: This MA57YW antibody has been reported for use in flow cytometric analysis. This MA57YW monoclonal antibody recognizes human B7-H7 on live and paraformaldehyde-fixed cells. Applications Tested: This MA57YW antibody has been pre-diluted and tested by flow cytometric analysis of the HDLM-2 cell line. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
B7-H7 (also known as HHLA2) is a membrane protein with three Ig-like domains (IgV-IgC-IgV), whereas other members of the B7 family generally have only two Ig domains (IgV-IgC). B7-H7 is somewhat more closely related to B7-H3 and B7-H4 and shares 10% to 18% amino acid identity and 23% to 33% similarity to B7 family members. B7-H7 protein is widely expressed in epithelium cells of the gallbladder, breast, kidney, colon, small intestines and placenta. In the immune system, HHLA2 protein is not constitutively expressed on human monocytes/macrophages. B7-H7 is widely expressed in cancers, especially in triple negative breast cancer.Specifications
| B7-H7 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| B7-H5; B7H7; B7-H7; B7y; HERV-H LTR-associating 2; HERV-H LTR-associating protein 2; HHLA2; Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| HHLA2 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| MA57YW | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| Mouse | |
| Human | |
| Q9UM44 | |
| 11148 | |
| B7-H7 recombinant protein. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| HHLA2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.