missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Antibody (C80/1608) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£489.00
Specifications
| Antigen | B7-1/CD80 |
|---|---|
| Clone | C80/1608 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human samples. It is validated for ELISA, CyTOF-ready.Specifications
| B7-1/CD80 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
| PBS with No Preservative | |
| 941 | |
| Recombinant human full-length CD80 protein | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| C80/1608 | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| CD80 | |
| IgG1 κ | |
| Protein A or G purified | |
| 60 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title