missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Antibody (C80/1608) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54307-100UG
This item is not returnable.
View return policy
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human samples. It is validated for ELISA, CyTOF-ready.
Specifications
| B7-1/CD80 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| Mouse | |
| 60 kDa | |
| 100 μg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
| 941 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| C80/1608 | |
| Unconjugated | |
| PBS with No Preservative | |
| CD80 | |
| Recombinant human full-length CD80 protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction