missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALNT2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | B4GALNT2 |
|---|---|
| Concentration | 0.45 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16862615
|
Proteintech
24024-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16852615
|
Proteintech
24024-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody detects the 55-65 kDa B4GALNT2 protein.
Involved in the synthesis of the Sd(a) antigen (Sia-alpha2,3-[GalNAc-beta1,4]Gal-beta1,4-GlcNAc), a carbohydrate determit expressed on erythrocytes, the colonic mucosa and other tissues. Transfers a beta-1,4-linked GalNAc to the galactose residue of an alpha-2,3-sialylated chain.Specifications
| B4GALNT2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8NHY0 | |
| 124872 | |
| B4GALNT2 Fusion Protein Ag21166 | |
| Primary | |
| -20°C | |
| B4GALNT2 |
| 0.45 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B4GALGT2, B4GALNT2, B4GALT, Cad, GALGT2, SD, Sda | |
| B4GALNT2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title