missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALNT2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24024-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody detects the 55-65 kDa B4GALNT2 protein.
Involved in the synthesis of the Sd(a) antigen (Sia-alpha2,3-[GalNAc-beta1,4]Gal-beta1,4-GlcNAc), a carbohydrate determit expressed on erythrocytes, the colonic mucosa and other tissues. Transfers a beta-1,4-linked GalNAc to the galactose residue of an alpha-2,3-sialylated chain.Specifications
| B4GALNT2 | |
| Polyclonal | |
| Unconjugated | |
| B4GALNT2 | |
| B4GALGT2, B4GALNT2, B4GALT, Cad, GALGT2, SD, Sda | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124872 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NHY0 | |
| B4GALNT2 | |
| B4GALNT2 Fusion Protein Ag21166 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction