missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALNT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | B3GALNT1 |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16841595
|
Proteintech
22048-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16831595
|
Proteintech
22048-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B3GALNT1 gene ontology annotations related to this gene include protein glycosylation.Specifications
| B3GALNT1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| O75752, Q6AY39, Q920V1 | |
| 26879, 310508, 8706 | |
| B3GALNT1 Fusion Protein Ag16976 | |
| Primary | |
| -20°C | |
| B3GALNT1 |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| b3Gal T3, B3GALNT1, B3GALT3, Beta 1,3 GalNAc T1, Beta 1,3 GalTase 3, Beta 3 Gx T3, beta3Gal T3, Beta3GalT3, galT3, Gb4Cer, GLCT3, GLOB, Globoside synthase, P, P1 | |
| B3GALNT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title