missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALNT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22048-1-AP-150UL
This item is not returnable.
View return policy
Description
B3GALNT1 gene ontology annotations related to this gene include protein glycosylation.Specifications
| B3GALNT1 | |
| Polyclonal | |
| Unconjugated | |
| B3GALNT1 | |
| b3Gal T3, B3GALNT1, B3GALT3, Beta 1,3 GalNAc T1, Beta 1,3 GalTase 3, Beta 3 Gx T3, beta3Gal T3, Beta3GalT3, galT3, Gb4Cer, GLCT3, GLOB, Globoside synthase, P, P1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26879, 310508, 8706 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75752, Q6AY39, Q920V1 | |
| B3GALNT1 | |
| B3GALNT1 Fusion Protein Ag16976 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction