Learn More
Fisherbrand™ B24 Cone Adapter
Shop All Fisherbrand Products
Click to view available options
Material:
Rubber
Description
- Cone/rubber tubing
- Straight connection
- Cone size: B24
- GL14 thread size
Specifications
Specifications
| Material | Rubber |
| Product Type | Adapter |
| Standard Taper Joint No. | B24 |
| Thread Size | GL14 |