missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B23/NPM1 Mouse anti-Human, Mouse, Rat, CoraLite 532, Clone: 4F12A3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | B23/NPM1 |
|---|---|
| Clone | 4F12A3 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 532 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17731339
|
Proteintech
CL532-60096-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17609896
|
Proteintech
CL532-60096-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B23/NPM1 Monoclonal antibody apecifically detetects B23/NPM1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| B23/NPM1 | |
| Immunofluorescence | |
| CoraLite 532 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 4869 | |
| B23/NPM1 fusion protein Ag7415 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 4F12A3 | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| BC002398 | |
| NPM1 | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title