missing translation for 'onlineSavingsMsg'
Learn More

B23/NPM1 Mouse anti-Human, Mouse, Rat, CoraLite 532, Clone: 4F12A3, Proteintech

Product Code. 17731339 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17731339 20 μL 20µL
17609896 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17731339 Supplier Proteintech Supplier No. CL5326009620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

B23/NPM1 Monoclonal antibody apecifically detetects B23/NPM1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.

Specifications

Antigen B23/NPM1
Applications Immunofluorescence
Classification Monoclonal
Clone 4F12A3
Concentration 1000 ug/mL
Conjugate CoraLite 532
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC002398
Gene Symbols NPM1
Host Species Mouse
Immunogen B23/NPM1 fusion protein Ag7415
Purification Method Protein G purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 4869
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.