missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZI2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33293-100ul
This item is not returnable.
View return policy
Description
AZI2 Monoclonal antibody specifically detects AZI2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| AZI2 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| 5-azacytidine induced 2, AZ25-azacytidine-induced protein 2, FLJ21939, NAP15-azacytidine induced gene 2, NF-kappa-B-activating kinase-associated protein 1, TILPNak-associated protein 1 | |
| Recombinant protein containing a sequence corresponding to amino acids 282-387 of human AZI2 (NP_071906.1).,, Sequence:, YTRHPPLLPNGKALCHTTSSPLPGDVKVLSEKAILQSWTDNERSIPNDGTCFQEHSSYGRNSLEDNSWVFPSPPKSSETAFGETKTKTLPLPNLPPLHYLDQHNQN | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64343 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction