missing translation for 'onlineSavingsMsg'
Learn More

AY 9944 Dihydrochloride, Thermo Scientific Chemicals

Product Code. p-7082444
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 15486709

Brand: Thermo Scientific Alfa Aesar J65079.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibitor of hedgehog (Hh) signaling

Inhibitor of hedgehog (Hh) signaling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-93-8
Molecular Formula C22H28Cl2N2·2HCl, C22H30Cl4N2
Synonym 1,4-Bis(2-chlorobenzylaminomethyl)cyclohexane dihydrochloride

Specifications

Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) Inhibitor of hedgehog (Hh) signaling
Formula Weight 464.30g/mol
Sensitivity Hygroscopic
Product Type Signal Transduction Reagent
Form Solid
InChI Key NRVIEWRSGDDWHP-UHFFFAOYSA-N
SMILES [H+].[H+].[Cl-].[Cl-].ClC1=CC=CC=C1CNCC1CCC(CNCC2=CC=CC=C2Cl)CC1
IUPAC Name dihydrogen [(2-chlorophenyl)methyl]({[4-({[(2-chlorophenyl)methyl]amino}methyl)cyclohexyl]methyl})amine dichloride
Molecular Weight (g/mol) 464.30
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.